accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFOCl

CAS RN 51336-94-8

IUPAC Name: 2-chloro-1-(2,4-difluorophenyl)ethan-1-one
Synonyms: 2-chloro-1-(2,4-difluorophenyl)ethanone
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: UENGBOCGGKLVJJ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C(=O)CCl)C(F)=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results