accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51364-51-3

IUPAC Name: tris(1,5-diphenylpenta-1,4-dien-3-one) dipalladium
Synonyms: tris(dibenzylideneacetone)dipalladium(0)
Molecular Weight (g/mol): 915.73
Molecular Formula: C51H42O3Pd2
InChi Key: CYPYTURSJDMMMP-UHFFFAOYSA-N
SMILES: [Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results