accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51373-68-3

OOOOOOOOOOOOH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OYbYb

CAS RN 51373-68-3

IUPAC Name: diytterbium(3+) decahydrate trioxalate
Synonyms: diytterbium(3+) decahydrate trioxalate
Molecular Weight (g/mol): 790.31
Molecular Formula: C6H20O22Yb2
InChi Key: FUUFMJYYMRCJSY-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results