CAS RN 51554-07-5
CAS RN 51554-07-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycidyl Linolenate, TRC
CAS: 51554-07-5 Molecular Formula: C21 H34 O3 Molecular Weight (g/mol): 334.49 Synonym: (9Z,12Z,15Z)-9,12,15-Octadecatrienoic Acid 2-Oxiranylmethyl Ester; IUPAC Name: oxiran-2-ylmethyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate SMILES: CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OCC1CO1