missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Linolenate, TRC

Product Code. 30137463
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137463 500 mg 500mg
30007473 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137463 Supplier Toronto Research Chemicals Supplier No. TRCG615710500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51554-07-5
Molecular Formula C21 H34 O3
Molecular Weight (g/mol) 334.49
Synonym (9Z,12Z,15Z)-9,12,15-Octadecatrienoic Acid 2-Oxiranylmethyl Ester;
IUPAC Name oxiran-2-ylmethyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OCC1CO1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h3-4,6-7,9-10,20H,2,5,8,11-19H2,1H3/b4-3-,7-6-,10-9-
Formula Weight 334.2508
Chemical Name or Material Glycidyl Linolenate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.