accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSHOH

CAS RN 51755-83-0

IUPAC Name: 3-sulfanylhexan-1-ol
Synonyms: 3-sulfanylhexan-1-ol
Molecular Weight (g/mol): 134.24
Molecular Formula: C6H14OS
InChi Key: TYZFMFVWHZKYSE-UHFFFAOYNA-N
SMILES: CCCC(S)CCO
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)