accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OON

CAS RN 5176-27-2

IUPAC Name: tert-butyl 1H-pyrrole-1-carboxylate
Synonyms: tert-butyl pyrrole-1-carboxylate
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: IZPYBIJFRFWRPR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)