missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-1-carboxylic Acid tert-Butyl Ester, TRC

Product Code. 30411831
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411831 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411831 Supplier Toronto Research Chemicals Supplier No. TRCP99790050G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrrole-1-carboxylic Acid tert-Butyl Ester
CAS 5176-27-2
Molecular Formula C9H13NO2
Quantity 50 g
Synonym 1-(tert-Butoxycarbonyl)-1H-pyrrole, 1-(tert-Butoxycarbonyl)pyrrole, 1-Pyrrolecarboxylic Acid tert-Butyl Ester, N-(tert-Butyloxycarbonyl)-1H-pyrrole, N-(tert-Butyloxycarbonyl)pyrrole, N-BOC-pyrrole, N-tert-Butoxycarbonyl-1H-pyrrole, N-tert-Butoxycarbonylpyrrole, tert-Butyl 1H-Pyrrole-1-carboxylate, tert-Butyl Pyrrole-1-carboxylate, 1H-Pyrrole-1-carboxylic Acid','1,1-Dimethylethyl ester
InChI Formula InChI=1S/C9H13NO2/c1-9(2,3)12-8(11)10-6-4-5-7-10/h4-7H,1-3H3
SMILES O=C(N1C=CC=C1)OC(C)(C)C
IUPAC Name tert-butyl 1H-pyrrole-1-carboxylate
Molecular Weight (g/mol) 167.21
Formula Weight 167.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.