accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51773-92-3

IUPAC Name: hydrogen [2,8-bis(trifluoromethyl)quinolin-4-yl](piperidin-2-yl)methanol chloride
Synonyms: hydrogen mefloquine chloride
Molecular Weight (g/mol): 414.78
Molecular Formula: C17H17ClF6N2O
InChi Key: WESWYMRNZNDGBX-UHFFFAOYNA-N
SMILES: [H+].[Cl-].OC(C1CCCCN1)C1=C2C=CC=C(C2=NC(=C1)C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Concentration 
  • (1)
Industry Type 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)