Learn More
Mefloquine Hydrochloride, Mikromol™
Description
Mefloquine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Mefloquine hydrochloride |
| CAS | 51773-92-3 |
| Analyte or Component Names | Mefloquine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H16 F6 N2 O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Mefloquine hydrochloride, (RS)-[2,8-Bis(trifluoromethyl)quinolin-4-yl][(2SR)-piperidin-2-yl]methanol hydrochloride |
| InChI Formula | InChI=1S/C17H16F6N2O.ClH/c18-16(19,20)11-5-3-4-9-10(15(26)12-6-1-2-7-24-12)8-13(17(21,22)23)25-14(9)11;/h3-5,8,12,15,24,26H,1-2,6-7H2;1H/t12-,15+;/m0./s1 |
| SMILES | Cl.O[C@@H]([C@@H]1CCCCN1)c2cc(nc3c(cccc23)C(F)(F)F)C(F)(F)F |
| IUPAC Name | (R)-[2,8-bis(trifluoromethyl)quinolin-4-yl]-[(2S)-piperidin-2-yl]methanol;hydrochloride |
| Show More |