accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSOH3C

CAS RN 51792-34-8

IUPAC Name: 3,4-dimethoxythiophene
Synonyms: 3,4-dimethoxythiophene
Molecular Weight (g/mol): 144.19
Molecular Formula: C6H8O2S
InChi Key: ZUDCKLVMBAXBIF-UHFFFAOYSA-N
SMILES: COC1=CSC=C1OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results