accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51997-51-4

IUPAC Name: 4-[(oxiran-2-yl)methoxy]-9H-carbazole
Synonyms: 4-(oxiran-2-ylmethoxy)-9H-carbazole
Molecular Weight (g/mol): 239.27
Molecular Formula: C15H13NO2
InChi Key: SVWKIGRDISDRLO-UHFFFAOYNA-N
SMILES: C(OC1=CC=CC2=C1C1=C(N2)C=CC=C1)C1CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results