missing translation for 'onlineSavingsMsg'
Learn More

4-(2,3-Epoxypropoxy)carbazole, TRC

Product Code. 30138552
Change view
Click to view available options
Quantity:
100 mg
10 g
25 g
Unit Size:
100mg
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138552 10 g 10g
30133076 25 g 25g
17993375 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138552 Supplier Toronto Research Chemicals Supplier No. TRCE58925010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51997-51-4
Molecular Formula C15 H13 N O2
Molecular Weight (g/mol) 239.27
Synonym 4-(2,3-Epoxypropoxy)carbazole
IUPAC Name 4-(oxiran-2-ylmethoxy)-9H-carbazole
SMILES C(Oc1cccc2[nH]c3ccccc3c12)C4CO4

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H13NO2/c1-2-5-12-11(4-1)15-13(16-12)6-3-7-14(15)18-9-10-8-17-10/h1-7,10,16H,8-9H2
Formula Weight 239.0946
Chemical Name or Material 4-(2,3-Epoxypropoxy)carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.