accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHOHO

CAS RN 520-36-5

IUPAC Name: 5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Synonyms: chamomile spigenin C.I. natural yellow 1 apigenine 7-trihydroxyflavone 4,5 versulin pelargidenon 1449 4,5, 7-trihydroxyflavone apigenol
Molecular Weight (g/mol): 270.24
Molecular Formula: C15H10O5
InChi Key: KZNIFHPLKGYRTM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Product Line 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)