missing translation for 'onlineSavingsMsg'
Learn More

Apigenin 100 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer

Product Code. 30317484
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317484 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317484 Supplier Dr. Ehrenstorfer Supplier No. DREA10290600MC100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Apigenin
CAS 520-36-5
Analyte or Component Names Apigenin
Shipping Condition Room Temperature
Molecular Formula C15 H10 O5
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',5,7-Trihydroxyflavone, Pelargidenon 1449;
InChI Formula InChI=1S/C15H10O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-7,16-18H
SMILES Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
Solvent or Matrix Acetonitrile/Methanol
Molecular Weight (g/mol) 270.24
Quantity 1 mL
Formula Weight 270.0528 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.