accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH2N

CAS RN 5267-34-5

IUPAC Name: 1,1-diphenylmethanamine hydrochloridyl
Synonyms: benzhydrylamin hydrochloridyl
Molecular Weight (g/mol): 218.7
Molecular Formula: C13H13ClN
InChi Key: WYZLZIDEQFCQPH-UHFFFAOYSA-N
SMILES: [Cl].NC(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results