CAS RN 52767-84-7
IUPAC Name:
5-propyl-1,2-dihydropyrimidine-2-thione
Synonyms:
5-propyl-1H-pyrimidine-2-thione
Molecular Weight (g/mol):
154.23
Molecular Formula:
C7H10N2S
InChi Key:
YZFGTZRDMRKBBU-UHFFFAOYSA-N
SMILES:
CCCC1=CNC(=S)N=C1
1
–
1
of
1
results
1
2-Mercapto-5-n-propylpyrimidine, 98%
CAS: 52767-84-7 Molecular Formula: C7H10N2S Molecular Weight (g/mol): 154.23 MDL Number: MFCD07777095 InChI Key: YZFGTZRDMRKBBU-UHFFFAOYSA-N Synonym: 5-propylpyrimidine-2-thiol,2-mercapto-5-n-propylpyrimidine,pubchem9576,5-propylpyrimidine-2 1h-thione PubChem CID: 12356156 SMILES: CCCC1=CNC(=S)N=C1