missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-n-propylpyrimidine, 98%

Product Code. 11362249
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362249 25 g 25g
11342249 1 g 1g
11352249 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362249 Supplier Thermo Scientific Alfa Aesar Supplier No. H27239.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52767-84-7 | C7H10N2S | 154.23 g/mol

By using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
By using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52767-84-7
Molecular Formula C7H10N2S
Molecular Weight (g/mol) 154.23
MDL Number MFCD07777095
InChI Key YZFGTZRDMRKBBU-UHFFFAOYSA-N
Synonym 5-propylpyrimidine-2-thiol, 2-mercapto-5-n-propylpyrimidine, pubchem9576, 5-propylpyrimidine-2 1h-thione
PubChem CID 12356156
SMILES CCCC1=CNC(=S)N=C1

Specifications

Melting Point 207°C to 211°C
Quantity 25 g
IUPAC Name 5-propyl-1,2-dihydropyrimidine-2-thione
Formula Weight 154.24
Percent Purity 98%
Chemical Name or Material 2-Mercapto-5-n-propylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.