accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52809-07-1

H2N(S)NOOHNOHOO

CAS RN 52809-07-1

IUPAC Name: (2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
Synonyms: quisqualate
Molecular Weight (g/mol): 189.13
Molecular Formula: C5H7N3O5
InChi Key: ASNFTDCKZKHJSW-REOHCLBHSA-N
SMILES: N[C@@H](CN1OC(=O)NC1=O)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)