missing translation for 'onlineSavingsMsg'
Learn More

(+)-Quisqualic Acid, TRC

Product Code. 30425631
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425631 5 mg 5mg
30418950 25 mg -
30420809 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425631 Supplier Toronto Research Chemicals Supplier No. TRCQ8250005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52809-07-1
Molecular Formula C5 H7 N3 O5
Molecular Weight (g/mol) 189.13
Synonym L-Quisqualic acid
IUPAC Name (2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
SMILES N[C@@H](CN1OC(=O)NC1=O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H7N3O5/c6-2(3(9)10)1-8-4(11)7-5(12)13-8/h2H,1,6H2,(H,9,10)(H,7,11,12)/t2-/m0/s1
Formula Weight 189.0386
Chemical Name or Material (+)-Quisqualic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.