accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5289-74-7

H3CH3COH(R)OH(R)CH3HO(S)(S)OHO(R)(R)OH(S)OH(R)H3C(R)(R)H3C

CAS RN 5289-74-7

IUPAC Name: (1S,3aS,5aR,7R,8S,9aR,9bR,11aR)-3a,7,8-trihydroxy-9a,11a-dimethyl-1-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-5-one
Synonyms: the-7 crustecdysone crustecdyson polypodine A β ecdysone β-ecdysone isoinokosterone commisterone viticosterone 20 hydroxyecdysone
Molecular Weight (g/mol): 480.64
Molecular Formula: C27H44O7
InChi Key: NKDFYOWSKOHCCO-YPVLXUMRSA-N
SMILES: CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)