CAS RN 53078-70-9
IUPAC Name:
2-methoxy-5-methylbenzonitrile
Synonyms:
2-methoxy-5-methylbenzonitrile
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
MYOASWSDRGHGLN-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C)C=C1C#N
1
–
1
of
1
results
1
2-Methoxy-5-methylbenzonitrile, 97%, Thermo Scientific™
CAS: 53078-70-9 Molecular Formula: C9H9NO Molecular Weight (g/mol): 147.18 InChI Key: MYOASWSDRGHGLN-UHFFFAOYSA-N Synonym: 2-methoxy-5-methyl-benzonitrile,acmc-1an2t,benzonitrile,2-methoxy-5-methyl,2-methoxy-5-methylbenzonitrile PubChem CID: 3801023 IUPAC Name: 2-methoxy-5-methylbenzonitrile SMILES: CC1=CC(=C(C=C1)OC)C#N