accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 533-67-5

IUPAC Name: (2S,4S,5R)-oxane-2,4,5-triol
Synonyms: D-2-deoxyribose thyminose
Molecular Weight (g/mol): 134.13
Molecular Formula: C5H10O4
InChi Key: ZVQAVWAHRUNNPG-LMVFSUKVSA-N
SMILES: O[C@@H]1C[C@H](O)[C@H](O)CO1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)