accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5362-56-1

IUPAC Name: (2Z)-4-methylpent-2-enal
Synonyms: (2Z)-4-methylpent-2-enal
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: RIWPMNBTULNXOH-ARJAWSKDSA-N
SMILES: CC(C)\C=C/C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results