accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH2NOOCH3

CAS RN 5372-81-6

IUPAC Name: 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate
Synonyms: dimethyl-2-amino-P-phthalate
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: DSSKDXUDARIMTR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C(=O)OC)C(N)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results