accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3(E/Z)

CAS RN 53744-28-8

IUPAC Name: 3-(3,4-dimethoxyphenyl)-1-phenylprop-2-en-1-one
Synonyms: 3-(3,4-dimethoxyphenyl)-1-phenylprop-2-en-1-one
Molecular Weight (g/mol): 268.31
Molecular Formula: C17H16O3
InChi Key: LSHZPTCZLWATBZ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C=CC(=O)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results