accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ON

CAS RN 53882-81-8

IUPAC Name: 3-(3-methylphenyl)-3-oxopropanenitrile
Synonyms: 3-(3-methylphenyl)-3-oxopropanenitrile
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: IVLKDYOTZMFMLO-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)CC#N
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results