CAS RN 53911-68-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-(4-Chlorophenyl)glutaric Anhydride, TRC
CAS: 53911-68-5 Molecular Formula: C11 H9 Cl O3 Molecular Weight (g/mol): 224.64 Synonym: 4-(4-chlorophenyl)dihydro-2H-pyran-2,6(3H)-dione,4-(4-Chlorophenyl)tetrahydropyran-2,6-dione, 3-(4-Chlorophenyl)glutaric Anhydride IUPAC Name: 4-(4-chlorophenyl)oxane-2,6-dione SMILES: Clc1ccc(cc1)C2CC(=O)OC(=O)C2
2
4-(4-Chlorophenyl)tetrahydropyran-2,6-dione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.