missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenyl)glutaric Anhydride, TRC

Product Code. 30135313
Change view
Click to view available options
Quantity:
250 mg
500 mg
5 g
Unit Size:
250mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135313 500 mg 500mg
30134878 5 g 5g
17954025 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135313 Supplier Toronto Research Chemicals Supplier No. TRCC378300500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53911-68-5
Molecular Formula C11 H9 Cl O3
Molecular Weight (g/mol) 224.64
Synonym 4-(4-chlorophenyl)dihydro-2H-pyran-2,6(3H)-dione, 4-(4-Chlorophenyl)tetrahydropyran-2,6-dione, 3-(4-Chlorophenyl)glutaric Anhydride
IUPAC Name 4-(4-chlorophenyl)oxane-2,6-dione
SMILES Clc1ccc(cc1)C2CC(=O)OC(=O)C2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H9ClO3/c12-9-3-1-7(2-4-9)8-5-10(13)15-11(14)6-8/h1-4,8H,5-6H2
Formula Weight 224.024
Chemical Name or Material 3-(4-Chlorophenyl)glutaric Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.