CAS RN 5392-10-9
IUPAC Name:
2-bromo-4,5-dimethoxybenzaldehyde
Synonyms:
2-bromo-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol):
245.07
Molecular Formula:
C9H9BrO3
InChi Key:
UQQROBHFUDBOOK-UHFFFAOYSA-N
SMILES:
COC1=CC(Br)=C(C=O)C=C1OC
1
–
1
of
1
results
1
6-Bromoveratraldehyde, 97%
CAS: 5392-10-9 Molecular Formula: C9H9BrO3 Molecular Weight (g/mol): 245.07 MDL Number: MFCD00003301 InChI Key: UQQROBHFUDBOOK-UHFFFAOYSA-N