accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5398-11-8

OO

CAS RN 5398-11-8

IUPAC Name: 3-phenyl-1,3-dihydro-2-benzofuran-1-one
Synonyms: 3-phenyl-3H-2-benzofuran-1-one
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: SQFMIHCARVMICF-UHFFFAOYNA-N
SMILES: O=C1OC(C2=CC=CC=C12)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results