accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3COOOOCH3CH3H3C

CAS RN 541-16-2

IUPAC Name: 1,3-di-tert-butyl propanedioate
Synonyms: 1,3-di-tert-butyl propanedioate
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: CLPHAYNBNTVRDI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CC(=O)OC(C)(C)C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)