accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHO

CAS RN 541-57-1

IUPAC Name: 5-hydroxy-2,3-dihydrofuran-3-one
Synonyms: 5-hydroxy-2H-furan-3-one
Molecular Weight (g/mol): 100.07
Molecular Formula: C4H4O3
InChi Key: YQCKTPIJASVZFF-UHFFFAOYSA-N
SMILES: OC1=CC(=O)CO1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results