accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5411-12-1

IUPAC Name: 2-benzoyl-3-phenyloxirane
Synonyms: 2-benzoyl-3-phenyloxirane
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: UQGMJZQVDNZRKT-UHFFFAOYNA-N
SMILES: O=C(C1OC1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results