accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrOBr

CAS RN 5414-19-7

IUPAC Name: 1-bromo-2-(2-bromoethoxy)ethane
Synonyms: 1-bromo-2-(2-bromoethoxy)ethane
Molecular Weight (g/mol): 231.92
Molecular Formula: C4H8Br2O
InChi Key: FOZVXADQAHVUSV-UHFFFAOYSA-N
SMILES: BrCCOCCBr
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)