CAS RN 54221-96-4
IUPAC Name:
6-methoxypyridine-2-carbaldehyde
Synonyms:
6-methoxypyridine-2-carbaldehyde
Molecular Weight (g/mol):
137.14
Molecular Formula:
C7H7NO2
InChi Key:
YDNWTNODZDSPNZ-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(C=O)=N1
1
–
1
of
1
results
1
6-Methoxypyridine-2-carboxaldehyde, 96%
CAS: 54221-96-4 Molecular Formula: C7H7NO2 Molecular Weight (g/mol): 137.138 MDL Number: MFCD05664030 InChI Key: YDNWTNODZDSPNZ-UHFFFAOYSA-N Synonym: 6-methoxypicolinaldehyde,6-methoxy-2-pyridinecarboxaldehyde,2-methoxy-6-pyridinecarboxaldehyde,2-formyl-6-methoxypyridine,6-methoxy-pyridine-2-carbaldehyde,2-pyridinecarboxaldehyde, 6-methoxy,6-methoxypyridine-2-carboxaldehyde,pubchem15345,acmc-1aulb,ksc490s1l PubChem CID: 12325392 IUPAC Name: 6-methoxypyridine-2-carbaldehyde SMILES: COC1=CC=CC(=N1)C=O