accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrOSN

CAS RN 54223-20-0

IUPAC Name: 1-(1,3-benzothiazol-2-yl)-2-bromoethan-1-one
Synonyms: 1-(1,3-benzothiazol-2-yl)-2-bromoethanone
Molecular Weight (g/mol): 256.12
Molecular Formula: C9H6BrNOS
InChi Key: AYWGYNKWZWBMSV-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=NC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results