accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OHHC

CAS RN 5430-01-3

IUPAC Name: 3-methylnon-1-yn-3-ol
Synonyms: 3-methylnon-1-yn-3-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: VQUXVWMAXIQKTQ-UHFFFAOYNA-N
SMILES: CCCCCCC(C)(O)C#C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results