accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5437-45-6

IUPAC Name: benzyl 2-bromoacetate
Synonyms: benzyl 2-bromoacetate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: JHVLLYQQQYIWKX-UHFFFAOYSA-N
SMILES: BrCC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)