missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetic Acid Phenylmethyl Ester, TRC

Product Code. 30330596
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330596 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330596 Supplier Toronto Research Chemicals Supplier No. TRCB67911550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoacetic Acid Phenylmethyl Ester
CAS 5437-45-6
Molecular Formula C9H9BrO2
Quantity 50 g
Synonym Bromoacetic Acid Benzyl Ester, 2-Bromoacetic Acid Benzyl Ester, Benzyl 2-Bromoacetate, Benzyl Bromoacetate, Bromoacetic Acid Benzyl Ester, Merbac 35, NSC 16114, NSC 23980, Phenylmethyl Bromoacetate
InChI Formula InChI=1S/C9H9BrO2/c10-6-9(11)12-7-8-4-2-1-3-5-8/h1-5H,6-7H2
SMILES BrCC(=O)OCc1ccccc1
IUPAC Name benzyl 2-bromoacetate
Molecular Weight (g/mol) 229.07
Formula Weight 227.9786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.