accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5466-06-8

IUPAC Name: ethyl 3-sulfanylpropanoate
Synonyms: ethyl 3-sulfanylpropanoate
Molecular Weight (g/mol): 134.19
Molecular Formula: C5H10O2S
InChi Key: CJQWLNNCQIHKHP-UHFFFAOYSA-N
SMILES: CCOC(=O)CCS
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results