missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Mercaptopropanoate, TRC

Product Code. 30382585
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382585 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382585 Supplier Toronto Research Chemicals Supplier No. TRCE932340100G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3-Mercaptopropanoate
CAS 5466-06-8
Molecular Formula C5H10O2S
Quantity 100 g
Synonym 3-Mercapto-propanoic Acid Ethyl Ester, 3-Mercapto-propionic Acid Ethyl Ester, 3-Mercaptopropanoic Acid Ethyl Ester, 3-Mercaptopropionic Acid Ethyl Ester, EPM, EPM (thiol), Ethyl 3-Mercaptopropanoate, Ethyl 3-Mercaptopropionate, Ethyl beta-Mercaptopropionate, NSC 25754, beta-Mercaptopropionic Acid Ethyl Ester
InChI Formula InChI=1S/C5H10O2S/c1-2-7-5(6)3-4-8/h8H,2-4H2,1H3
SMILES CCOC(=O)CCS
IUPAC Name ethyl 3-sulfanylpropanoate
Molecular Weight (g/mol) 134.2
Formula Weight 134.0402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.