accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HOHCl

CAS RN 5470-11-1

IUPAC Name: hydrogen hydroxylamine chloride
Synonyms: hydroxylammonium chloride
Molecular Weight (g/mol): 69.49
Molecular Formula: ClH4NO
InChi Key: WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NO
Quantity 
  • (7)
  • (10)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (11)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (4)
  • (4)
  • (4)
  • (9)
Physical Form 
  • (11)
  • (1)
  • (1)
  • (8)
Grade 
  • (3)
  • (3)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (6)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (11)
  • (7)
  • (1)
  • (7)
  • (10)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (11)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (4)
  • (9)
  • (11)
  • (1)
  • (1)
  • (8)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)