accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOON

CAS RN 54950-20-8

IUPAC Name: methyl 3-oxo-3-(pyridin-3-yl)propanoate
Synonyms: methyl 3-oxo-3-(pyridin-3-yl)propanoate
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: JUQKVXRLRKKRPL-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C1=CC=CN=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results