CAS RN 55443-55-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Glutamic Acid-13C5, TRC
CAS: 55443-55-5 Molecular Formula: 13C5H9NO4 Molecular Weight (g/mol): 152.09 Synonym: L-Glutamic-1,2,3,4,5-13C5 Acid SMILES: N[13C@@H]([13CH2][13CH2][13C](O)=O)[13C](O)=O