accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH2N

CAS RN 5586-73-2

IUPAC Name: hydrogen 3,3-diphenylpropan-1-amine chloride
Synonyms: 3,3-diphenylpropylamine
Molecular Weight (g/mol): 247.77
Molecular Formula: C15H18ClN
InChi Key: AHHLKJBLNXAJID-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)