accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

TiH3COCH3OCH3OCH3O

CAS RN 5593-70-4

IUPAC Name: titanium(4+) tetrakis(butan-1-olate)
Synonyms: titanium butoxide
Molecular Weight (g/mol): 340.33
Molecular Formula: C16H36O4Ti
InChi Key: YHWCPXVTRSHPNY-UHFFFAOYSA-N
SMILES: [Ti+4].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-]
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results