accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHNOClClOHNOO

CAS RN 56-75-7

IUPAC Name: 2,2-dichloro-N-[1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
Synonyms: cloranfenicol
Molecular Weight (g/mol): 323.13
Molecular Formula: C11H12Cl2N2O5
InChi Key: WIIZWVCIJKGZOK-UHFFFAOYNA-N
SMILES: OCC(NC(=O)C(Cl)Cl)C(O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (11)
  • (5)
Physical Form 
  • (4)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Type 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (11)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)