accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBrOO

CAS RN 56073-10-0

IUPAC Name: 3-(3-{4'-bromo-[1,1'-biphenyl]-4-yl}-1,2,3,4-tetrahydronaphthalen-1-yl)-2-hydroxy-4H-chromen-4-one
Synonyms: brodifacoum
Molecular Weight (g/mol): 523.43
Molecular Formula: C31H23BrO3
InChi Key: LNDUTAIPEYOCDF-UHFFFAOYNA-N
SMILES: OC1=C(C2CC(CC3=CC=CC=C23)C2=CC=C(C=C2)C2=CC=C(Br)C=C2)C(=O)C2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results