missing translation for 'onlineSavingsMsg'
Learn More

Rodenticides Mixture 248 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313137
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313137 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313137 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000248AL
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brodifacoum∣Bromadiolone∣Chlorophacinone∣Difenacoum∣Diphacinone∣Warfarin
CAS 56073-10-0, 28772-56-7, 3691-35-8, 56073-07-5, 82-66-6, 81-81-2
Analyte or Component Names Brodifacoum∣Bromadiolone∣Chlorophacinone∣Difenacoum∣Diphacinone∣Warfarin
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Brodifacoum 100 μg/mL∣Bromadiolone 100 μg/mL∣Chlorophacinone 100 μg/mL∣Difenacoum 100 μg/mL∣Diphacinone 100 μg/mL∣Warfarin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.